siRNA Details
| Gene Symbol: Pcsk7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281934.1 | NP_001268863.1 | 4 | 1395 | ||
| NM_008794.2 | NP_032820.2 | 4 | 1131 | |||
| GenBank mRNA | AK148416.1 | - | 4 | 716 | ||
| AK151980.1 | - | 4 | 718 | |||
| AK165389.1 | - | 4 | 737 | |||
| BC006730.1 | - | 3 | 632 | 3962441 | ||
| BC007322.1 | - | 4 | 994 | |||
| BC056456.1 | - | 4 | 1131 | |||
| U48830.1 | - | 4 | 703 | |||

Pcsk7
Genomic Map