siRNA Details
| Gene Symbol: Pcsk7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281934.1 | NP_001268863.1 | 17 | 3524 | ||
| NM_008794.2 | NP_032820.2 | 17 | 3260 | |||
| GenBank mRNA | AK148416.1 | - | 17 | 2845 | ||
| AK151980.1 | - | 17 | 2847 | |||
| AK210461.1 | - | 1 | 207 | |||
| BC006730.1 | - | 16 | 2761 | 3962441 | ||
| BC007322.1 | - | 17 | 3121 | |||
| BC056456.1 | - | 18 | 3334 | |||
| U48830.1 | - | 17 | 2832 | |||

Pcsk7
Genomic Map