siRNA Details
| Gene Symbol: Pcx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162946.1 | NP_001156418.1 | 21 | 3473 | ||
| NM_008797.3 | NP_032823.2 | 20 | 3450 | |||
| GenBank mRNA | AK077665.1 | - | 12 | 2278 | ||
| AK148328.1 | - | 21 | 3420 | |||
| AK170596.1 | - | 21 | 3433 | |||
| AK170871.1 | - | 21 | 3424 | |||
| AK172320.1 | - | 21 | 3401 | |||
| BC055030.1 | - | 21 | 3408 | 5053475 | ||
| L09192.1 | - | 20 | 3426 | |||
| M97957.1 | - | 15 | 2598 | |||

Pcx
Genomic Map