siRNA Details
Gene Symbol: Pdcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008798.2 | NP_032824.1 | 5 | 1397 | ||
GenBank mRNA | AK039828.1 | - | 5 | 1377 | ||
X67914.1 | - | 5 | 1397 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pdcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008798.2 | NP_032824.1 | 5 | 1397 | ||
GenBank mRNA | AK039828.1 | - | 5 | 1377 | ||
X67914.1 | - | 5 | 1397 |