siRNA Details
| Gene Symbol: Pdha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008810.2 | NP_032836.1 | 11 | 1118 | ||
| GenBank mRNA | AK145718.1 | - | 11 | 1196 | ||
| AK148459.1 | - | 11 | 1139 | |||
| AK167987.1 | - | 11 | 1144 | |||
| AK168152.1 | - | 11 | 1168 | |||
| AK169289.1 | - | 11 | 1193 | |||
| AK187913.1 | - | 2 | 191 | |||
| BC007142.1 | - | 11 | 1118 | 3600605 | ||
| M76727.1 | - | 11 | 1205 | |||

Pdha1
Genomic Map