siRNA Details
| Gene Symbol: Pdha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008810.2 | NP_032836.1 | 11 | 2105 | ||
| GenBank mRNA | AK145718.1 | - | 11 | 2183 | ||
| AK148459.1 | - | 11 | 2126 | |||
| AK168152.1 | - | 11 | 2156 | |||
| AK169289.1 | - | 11 | 2180 | |||
| AL355693.1 | - | 1 | 23 | |||
| BC007142.1 | - | 11 | 2105 | 3600605 | ||

Pdha1
Genomic Map