siRNA Details
| Gene Symbol: Rhox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008818.2 | NP_032844.2 | 2, 3 | 567 | ||
| GenBank mRNA | AK146379.1 | - | 2, 3 | 592 | ||
| AK146393.1 | - | 2, 3 | 591 | |||
| AK146449.1 | - | 2, 3 | 573 | |||
| BC056204.1 | - | 3, 4 | 575 | 30022528 | ||
| DQ058642.1 | - | 2, 3 | 551 | |||
| M32484.1 | - | 2, 3 | 551 | |||

Rhox5
Genomic Map