siRNA Details
| Gene Symbol: Pet2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008821.2 | NP_032847.2 | 12 | 3581 | ||
| GenBank mRNA | BC138080.1 | - | 11 | 3402 | 8861099 | |
| M95564.1 | - | 12 | 3586 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pet2| Gene Symbol: Pet2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008821.2 | NP_032847.2 | 12 | 3581 | ||
| GenBank mRNA | BC138080.1 | - | 11 | 3402 | 8861099 | |
| M95564.1 | - | 12 | 3586 | |||