siRNA Details
| Gene Symbol: Pex7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008822.2 | NP_032848.1 | 3, 4 | 433 | ||
| GenBank mRNA | AK004687.1 | - | 3, 4 | 418 | ||
| AK160195.1 | - | 3, 4 | 397 | |||
| BC132213.1 | - | 3, 4 | 406 | 40130490 | ||
| BC132503.1 | - | 3, 4 | 406 | 40130780 | ||
| DV660555.1 | - | 3, 4 | 434 | |||
| U69171.1 | - | 3, 4 | 424 | |||

Pex7
Genomic Map