siRNA Details
| Gene Symbol: Invs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281977.1 | NP_001268906.1 | 5 | 787 | ||
| NM_001281978.1 | NP_001268907.1 | 5 | 787 | |||
| NM_010569.4 | NP_034699.3 | 4 | 671 | |||
| GenBank mRNA | AF034860.3 | - | 4 | 613 | ||
| AJ010902.1 | - | 4 | 612 | |||
| AK041671.1 | - | 4 | 606 | |||
| AK158766.1 | - | 5 | 746 | |||
| BC126929.1 | - | 5 | 676 | 40081431 | ||
| CK639743.1 | - | 1 | 150 | |||

Invs
Genomic Map