siRNA Details
| Gene Symbol: Irs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010571.3 | NP_034701.1 | 1 | 502 | ||
| GenBank mRNA | AF035948.1 | - | 1 | 308 | ||
| AK030311.1 | - | 1 | 504 | |||
| AK040947.1 | - | 1 | 487 | |||
| BC098235.1 | - | 1 | 334 | 1546952 | ||

Irs3
Genomic Map