siRNA Details
| Gene Symbol: Itih2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010582.3 | NP_034712.2 | 16 | 2203 | ||
| GenBank mRNA | AK133782.1 | - | 16 | 2102 | ||
| BC034341.1 | - | 16 | 2177 | 4211576 | ||
| X70392.1 | - | 16 | 2189 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itih2| Gene Symbol: Itih2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010582.3 | NP_034712.2 | 16 | 2203 | ||
| GenBank mRNA | AK133782.1 | - | 16 | 2102 | ||
| BC034341.1 | - | 16 | 2177 | 4211576 | ||
| X70392.1 | - | 16 | 2189 | |||