siRNA Details
| Gene Symbol: Itln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB016496.1 | - | 6 | 703 | ||
| AY157363.1 | - | 6 | 693 | |||
| BC150795.1 | - | 6 | 646 | 9087706 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itln1| Gene Symbol: Itln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB016496.1 | - | 6 | 703 | ||
| AY157363.1 | - | 6 | 693 | |||
| BC150795.1 | - | 6 | 646 | 9087706 | ||