siRNA Details
| Gene Symbol: Uhmk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010633.3 | NP_034763.3 | 3 | 841 | ||
| GenBank mRNA | AK030152.1 | - | 658 | |||
| AK218845.1 | - | 1 | 56 | |||
| BC058732.1 | - | 3 | 841 | 6414877 | ||
| X82320.1 | - | 2 | 327 | |||
| Y10725.2 | - | 3 | 698 | |||

Uhmk1
Genomic Map