siRNA Details
Gene Symbol: Fabp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272097.1 | NP_001259026.1 | 4 | 530 | ||
NM_001272098.1 | NP_001259027.1 | 3 | 360 | |||
NM_010634.3 | NP_034764.1 | 4 | 533 | |||
GenBank mRNA | AK008782.1 | - | 4 | 527 | ||
AK011551.1 | - | 4 | 529 | |||
AK166506.1 | - | 4 | 526 | |||
AK167389.1 | - | 4 | 528 | |||
BC002008.1 | - | 4 | 513 | 3490535 | ||
BC053042.1 | - | 2 | 3093 | |||
BC100543.1 | - | 4 | 508 | 30136900 | ||
W54445.1 | - | 3 | 313 | |||
X70100.1 | - | 4 | 512 |
