siRNA Details
| Gene Symbol: Klrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010654.3 | NP_034784.1 | 5 | 366 | ||
| GenBank mRNA | AF030311.1 | - | 4 | 346 | ||
| AF030312.1 | - | 5 | 395 | |||
| AF039025.1 | - | 5 | 360 | |||
| AF057714.1 | - | 4 | 346 | |||
| AK136548.1 | - | 5 | 367 | |||
| AK162976.1 | - | 6 | 427 | |||
| BC117112.1 | - | 5 | 384 | 40126431 | ||
| BC120854.1 | - | 5 | 386 | 40129777 | ||

Klrd1
Genomic Map