siRNA Details
| Gene Symbol: Sspn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010656.2 | NP_034786.1 | 3 | 2453 | ||
| GenBank mRNA | U02487.1 | - | 3 | 2454 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sspn| Gene Symbol: Sspn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010656.2 | NP_034786.1 | 3 | 2453 | ||
| GenBank mRNA | U02487.1 | - | 3 | 2454 | ||