siRNA Details
| Gene Symbol: Krt18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010664.2 | NP_034794.2 | 5 | 898 | ||
| GenBank mRNA | AK145413.1 | - | 5 | 888 | ||
| AK167072.1 | - | 5 | 887 | |||
| AK167265.1 | - | 5 | 882 | |||
| AK167432.1 | - | 5 | 885 | |||
| AK167469.1 | - | 5 | 888 | |||
| AK167676.1 | - | 5 | 887 | |||
| BC020474.1 | - | 5 | 888 | |||
| BC089022.1 | - | 5 | 860 | 30701486 | ||
| M11686.1 | - | 5 | 889 | |||
| M36376.1 | - | 5 | 884 | |||

Krt18
Genomic Map