siRNA Details
| Gene Symbol: Limk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK141813.1 | - | 16 | 2613 | ||
| AK170166.1 | - | 16 | 2939 | |||
| AK172083.1 | - | 15 | 2372 | |||
| BC063768.1 | - | 16 | 2593 | |||
| U14166.1 | - | 16 | 2635 | |||
| U15159.1 | - | 16 | 2623 | |||

Limk1
Genomic Map