siRNA Details
| Gene Symbol: Lipe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039507.2 | NP_001034596.1 | 2 | 289 | ||
| NM_010719.5 | NP_034849.2 | 2 | 725 | |||
| GenBank mRNA | AK019846.1 | - | 2 | 1126 | ||
| AK029984.1 | - | 2 | 728 | |||
| AK169858.1 | - | 2 | 284 | |||
| BC021642.1 | - | 2 | 285 | 4188567 | ||
| BC053502.1 | - | 2 | 349 | |||
| U08188.1 | - | 1 | 810 | |||

Lipe
Genomic Map