siRNA Details
| Gene Symbol: OMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005014.2 | NP_005005.1 | 3 | 1519 | ||
| GenBank mRNA | AB000114.1 | - | 3 | 1250 | ||
| AK225922.1 | - | 3 | 1364 | |||
| AY358872.1 | - | 3 | 1417 | |||
| BC046356.1 | - | 3 | 1421 | 5239983 | ||

OMD
Genomic Map