siRNA Details
| Gene Symbol: Lyn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111096.1 | NP_001104566.1 | 13 | 2914 | ||
| NM_010747.2 | NP_034877.2 | 13 | 2851 | |||
| GenBank mRNA | AK028112.1 | - | 13 | 2847 | ||
| AK152199.1 | - | 13 | 2913 | |||
| AK153038.1 | - | 13 | 2848 | |||
| BC031547.1 | - | 13 | 2885 | 3589207 | ||
| BC157998.1 | - | 13 | 2683 | 9007404 | ||

Lyn
Genomic Map