siRNA Details
| Gene Symbol: M6pr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010749.6 | NP_034879.2 | 7 | 1508 | ||
| GenBank mRNA | AK133943.1 | - | 7 | 1514 | ||
| AK149177.1 | - | 7 | 1538 | |||
| AK151607.1 | - | 7 | 1569 | |||
| AK153140.1 | - | 7 | 1519 | |||
| AK153302.1 | - | 7 | 1589 | |||
| AK153474.1 | - | 7 | 1529 | |||
| BC027210.1 | - | 7 | 1463 | |||
| BC046956.1 | - | 7 | 1508 | |||
| BC080811.1 | - | 7 | 1508 | 6438831 | ||
| M63286.1 | - | 7 | 1416 | |||
| X64068.1 | - | 7 | 1511 | |||

M6pr
Genomic Map