siRNA Details
| Gene Symbol: Matr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010771.6 | NP_034901.2 | 2 | 475 | ||
| GenBank mRNA | AK087939.1 | - | 2 | 360 | ||
| AK088351.1 | - | 2 | 368 | |||
| AK129199.1 | - | 2 | 426 | |||
| AK135937.1 | - | 1 | 208 | |||
| AK161128.1 | - | 2 | 374 | |||
| AK164188.1 | - | 1 | 253 | |||
| BC029070.1 | - | 2 | 386 | 3989914 | ||
| BC090833.1 | - | 1 | 248 | 6854095 | ||
| CT010322.1 | - | 1 | 159 | |||

Matr3
Genomic Map