siRNA Details
| Gene Symbol: Mlf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039543.2 | NP_001034632.1 | 8 | 1052 | ||
| NM_010801.3 | NP_034931.2 | 7 | 1007 | |||
| GenBank mRNA | AF009515.1 | - | 7 | 948 | ||
| AK133448.1 | - | 7 | 1008 | |||
| BC027526.1 | - | 8 | 1001 | 1495721 | ||

Mlf1
Genomic Map