siRNA Details
| Gene Symbol: Mnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010813.3 | NP_034943.3 | 6 | 3321 | ||
| GenBank mRNA | AK147933.1 | - | 6 | 3305 | ||
| BC054534.1 | - | 6 | 3247 | 5721249 | ||
| U77356.1 | - | 6 | 3318 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mnt| Gene Symbol: Mnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010813.3 | NP_034943.3 | 6 | 3321 | ||
| GenBank mRNA | AK147933.1 | - | 6 | 3305 | ||
| BC054534.1 | - | 6 | 3247 | 5721249 | ||
| U77356.1 | - | 6 | 3318 | |||