siRNA Details
| Gene Symbol: Morc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010816.1 | NP_034946.1 | 5 | 346 | ||
| GenBank mRNA | AF084945.1 | - | 5 | 346 | ||
| BC030893.1 | - | 5 | 295 | 4487929 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Morc1| Gene Symbol: Morc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010816.1 | NP_034946.1 | 5 | 346 | ||
| GenBank mRNA | AF084945.1 | - | 5 | 346 | ||
| BC030893.1 | - | 5 | 295 | 4487929 | ||