siRNA Details
| Gene Symbol: Msx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010836.3 | NP_034966.1 | 2 | 999 | ||
| GenBank mRNA | AK012208.1 | - | 2 | 998 | ||
| AK012556.1 | - | 2 | 1068 | |||
| BC051983.1 | - | 2 | 968 | |||
| BC137577.1 | - | 2 | 998 | 8860596 | ||
| BC137578.1 | - | 2 | 998 | 8860597 | ||
| U62523.1 | - | 2 | 1009 | |||

Msx3
Genomic Map