siRNA Details
| Gene Symbol: Map6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001043355.2 | NP_001036820.2 | 2 | 990 | ||
| NM_001048167.1 | NP_001041632.1 | 2 | 1889 | |||
| NM_010837.3 | NP_034967.2 | 2 | 1889 | |||
| GenBank mRNA | AK018193.2 | - | 2 | 1369 | ||
| AK087413.1 | - | 2 | 1852 | |||
| AK154455.1 | - | 2 | 962 | |||
| AK214985.1 | - | 2 | 292 | |||
| BC053039.1 | - | 2 | 1890 | 5709455 | ||
| BY259340.1 | - | 2 | 212 | |||
| Y16032.1 | - | 2 | 930 | |||

Map6
Genomic Map