siRNA Details
| Gene Symbol: Mtl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039657.2 | NP_001034746.1 | 9 | 1788 | ||
| NM_001286557.1 | NP_001273486.1 | 10 | 1872 | |||
| NM_001286558.1 | NP_001273487.1 | 9 | 1803 | |||
| GenBank mRNA | AB057423.1 | - | 9 | 1776 | ||
| AK161511.1 | - | 9 | 1789 | |||
| U67176.1 | - | 11 | 1805 | |||
| U77383.1 | - | 9 | 1425 | |||

Mtl5
Genomic Map