siRNA Details
| Gene Symbol: Myo5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010864.2 | NP_034994.2 | 4, 5 | 890 | ||
| GenBank mRNA | AK032713.1 | - | 4, 5 | 891 | ||
| AK149701.1 | - | 2, 3 | 334 | |||
| X57377.1 | - | 4, 5 | 503 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myo5a| Gene Symbol: Myo5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010864.2 | NP_034994.2 | 4, 5 | 890 | ||
| GenBank mRNA | AK032713.1 | - | 4, 5 | 891 | ||
| AK149701.1 | - | 2, 3 | 334 | |||
| X57377.1 | - | 4, 5 | 503 | |||