siRNA Details
| Gene Symbol: Myom1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083934.1 | NP_001077403.1 | 3 | 555 | ||
| NM_010867.2 | NP_034997.2 | 3 | 555 | |||
| GenBank mRNA | AJ012072.1 | - | 3 | 588 | ||
| AK052539.1 | - | 3 | 522 | |||
| AK143360.1 | - | 1945 | ||||
| BC060378.1 | - | 2 | 460 | 4980452 | ||
| CA460156.1 | - | 3 | 558 | |||
| Z22866.1 | - | 3 | 534 | |||

Myom1
Genomic Map