siRNA Details
| Gene Symbol: Ndp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC090623.1 | - | 3 | 1683 | 6807800 | |
| X92397.1 | - | 3 | 1528 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndp| Gene Symbol: Ndp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC090623.1 | - | 3 | 1683 | 6807800 | |
| X92397.1 | - | 3 | 1528 | |||