siRNA Details
| Gene Symbol: Nfe2l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010902.3 | NP_035032.1 | 5 | 1560 | ||
| GenBank mRNA | AK142347.1 | - | 5 | 1462 | ||
| AK144247.1 | - | 5 | 1434 | |||
| BC003340.1 | - | 5 | 1416 | |||
| BC026943.1 | - | 5 | 1501 | 3663276 | ||
| U20532.1 | - | 5 | 1576 | |||

Nfe2l2
Genomic Map