siRNA Details
| Gene Symbol: Nfs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010911.2 | NP_035041.2 | 2 | 232 | ||
| GenBank mRNA | AJ222660.1 | - | 2 | 231 | ||
| AK003786.1 | - | 2 | 226 | |||
| AK005724.1 | - | 2 | 221 | |||
| AK075575.1 | - | 2 | 230 | |||
| AK084831.1 | - | 2 | 201 | |||
| AK132838.1 | - | 2 | 233 | |||
| AK148527.1 | - | 2 | 233 | |||
| AK159828.1 | - | 2 | 221 | |||
| BC010586.1 | - | 2 | 210 | 3711352 | ||
| BC012651.1 | - | 2 | 218 | |||

Nfs1
Genomic Map