siRNA Details
| Gene Symbol: Nkx2-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010919.2 | NP_035049.1 | 2 | 1463 | ||
| GenBank mRNA | AK141002.1 | - | 2 | 1468 | ||
| BC138159.1 | - | 3 | 1284 | 8861178 | ||
| BC138160.1 | - | 3 | 1284 | 8861179 | ||
| U31566.1 | - | 2 | 1422 | |||

Nkx2-2
Genomic Map