siRNA Details
| Gene Symbol: Rrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010925.2 | NP_035055.2 | 13 | 1729 | ||
| GenBank mRNA | AF312394.1 | - | 13 | 1387 | ||
| AK077540.1 | - | 13 | 1654 | |||
| AK210980.1 | - | 1 | 226 | |||
| BC003913.1 | - | 6 | 825 | |||
| BC022910.1 | - | 12 | 1595 | 4165783 | ||
| BC065993.1 | - | 13 | 1691 | 5684672 | ||
| BC065994.1 | - | 13 | 1649 | 5685269 | ||
| BY616845.1 | - | 1 | 158 | |||
| U79773.1 | - | 14 | 1528 | |||
| U79774.1 | - | 13 | 1681 | |||

Rrp1
Genomic Map