siRNA Details
| Gene Symbol: Emc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010926.5 | NP_035056.1 | 2 | 390 | ||
| GenBank mRNA | AF052621.1 | - | 2 | 392 | ||
| AK042704.1 | - | 2 | 258 | |||
| AK076710.1 | - | 2 | 853 | |||
| AK143131.1 | - | 2 | 351 | |||
| AK148190.1 | - | 2 | 290 | |||
| AK161438.1 | - | 2 | 366 | |||
| AK164130.1 | - | 2 | 391 | |||
| AK201805.1 | - | 2 | 90 | |||
| BC009103.1 | - | 2 | 346 | 3482942 | ||

Emc8
Genomic Map