siRNA Details
| Gene Symbol: Emc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010926.5 | NP_035056.1 | 5 | 817 | ||
| GenBank mRNA | AF052621.1 | - | 5 | 819 | ||
| AK042704.1 | - | 5 | 685 | |||
| AK076710.1 | - | 7 | 1686 | |||
| AK143131.1 | - | 5 | 778 | |||
| AK148190.1 | - | 5 | 1328 | |||
| AK161438.1 | - | 5 | 793 | |||
| AK164130.1 | - | 5 | 818 | |||
| AK185099.1 | - | 1 | 19 | |||
| BC009103.1 | - | 5 | 773 | 3482942 | ||

Emc8
Genomic Map