siRNA Details
| Gene Symbol: Nr1i2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098404.1 | NP_001091874.1 | 5 | 907 | ||
| NM_010936.3 | NP_035066.1 | 5 | 1030 | |||
| GenBank mRNA | AF031814.1 | - | 5 | 845 | ||
| AK018630.1 | - | 5 | 1030 | |||
| BC120796.1 | - | 5 | 777 | 40129719 | ||
| BC137780.1 | - | 5 | 777 | 8860799 | ||

Nr1i2
Genomic Map