siRNA Details
| Gene Symbol: Nsg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010942.3 | NP_035072.2 | 5 | 1043 | ||
| GenBank mRNA | AF035683.1 | - | 5 | 992 | ||
| AK076451.1 | - | 5 | 1483 | |||
| AK078054.1 | - | 5 | 1034 | |||
| AK144503.1 | - | 5 | 1018 | |||
| AK148941.1 | - | 5 | 1043 | |||
| AX775765.1 | - | 5 | 1043 | |||
| BC008272.1 | - | 5 | 1029 | 3603440 | ||

Nsg1
Genomic Map