siRNA Details
| Gene Symbol: Oit3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010959.2 | NP_035089.1 | 7 | 1144 | ||
| GenBank mRNA | AF356506.1 | - | 7 | 1091 | ||
| AF517106.1 | - | 6 | 963 | |||
| AF517107.1 | - | 6 | 963 | |||
| AK040570.1 | - | 7 | 1145 | |||
| AK050081.1 | - | 7 | 1111 | |||
| AK153440.1 | - | 3 | 386 | |||
| AY180915.1 | - | 7 | 1068 | |||

Oit3
Genomic Map