siRNA Details
| Gene Symbol: Olfr49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010991.2 | NP_035121.1 | 1 | 306 | ||
| GenBank mRNA | BC145891.1 | - | 1 | 306 | 40131194 | |
| BC145893.1 | - | 1 | 306 | 40131196 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr49| Gene Symbol: Olfr49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010991.2 | NP_035121.1 | 1 | 306 | ||
| GenBank mRNA | BC145891.1 | - | 1 | 306 | 40131194 | |
| BC145893.1 | - | 1 | 306 | 40131196 | ||