siRNA Details
| Gene Symbol: Pax9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011041.2 | NP_035171.1 | 2 | 413 | ||
| GenBank mRNA | AK034985.1 | - | 2 | 410 | ||
| AK132404.1 | - | 2 | 414 | |||
| BC005794.1 | - | 2 | 353 | 3707718 | ||
| BC006930.1 | - | 1 | 1035 | |||
| X84000.1 | - | 2 | 385 | |||

Pax9
Genomic Map