siRNA Details
Gene Symbol: CDH18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291956.1 | NP_001278885.1 | 13, 14 | 2619 | ||
NM_004934.3 | NP_004925.1 | 11, 12 | 2162 | |||
XM_005248228.3 | XP_005248285.1 | 11, 12 | 2371 | |||
XM_006714435.3 | XP_006714498.1 | 10, 11 | 1978 | |||
XM_011513928.2 | XP_011512230.1 | 9, 10 | 1776 | |||
XM_011513929.2 | XP_011512231.1 | 8, 9 | 2030 | |||
XM_017008924.1 | XP_016864413.1 | 12, 13 | 2490 | |||
XM_017008925.1 | XP_016864414.1 | 12, 13 | 2486 | |||
XM_017008926.1 | XP_016864415.1 | 12, 13 | 2347 | |||
XM_017008927.1 | XP_016864416.1 | 12, 13 | 2276 | |||
XM_017008928.1 | XP_016864417.1 | 13, 14 | 2534 | |||
XM_017008929.1 | XP_016864418.1 | 10, 11 | 2014 | |||
XM_017008930.1 | XP_016864419.1 | 10, 11 | 1905 | |||
XM_017008931.1 | XP_016864420.1 | 11, 12 | 2158 | |||
XM_017008932.1 | XP_016864421.1 | 12, 13 | 2209 | |||
XM_017008933.1 | XP_016864422.1 | 12, 13 | 2338 | |||
XM_017008934.1 | XP_016864423.1 | 12, 13 | 2476 | |||
GenBank mRNA | AK289547.1 | - | 13, 14 | 2617 | ||
BC031051.1 | - | 11, 12 | 2118 | 5278717 | ||
U59325.1 | - | 10, 11 | 1941 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001081299.
Panther Classification:
Gene Ontology Categories:
