siRNA Details
| Gene Symbol: Cdk16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011049.4 | NP_035179.1 | 16 | 2047 | ||
| GenBank mRNA | AK051425.1 | - | 16 | 2048 | ||
| AK141083.1 | - | 10 | 1067 | |||
| AK145483.1 | - | 16 | 2077 | |||
| AK209172.1 | - | 1 | 193 | |||
| AK217385.1 | - | 1 | 193 | |||
| AK219069.1 | - | 1 | 193 | |||
| BC011069.1 | - | 16 | 1805 | 4006745 | ||
| X69025.1 | - | 16 | 2069 | |||

Cdk16
Genomic Map