siRNA Details
| Gene Symbol: Pdcd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168491.1 | NP_001161963.1 | 12 | 1913 | ||
| NM_011050.4 | NP_035180.2 | 12 | 2001 | |||
| GenBank mRNA | AK050175.1 | - | 1 | 132 | ||
| AK134366.1 | - | 12 | 2001 | |||
| AK172654.1 | - | 12 | 1866 | |||
| D50465.1 | - | 11 | 1774 | |||
| D86344.1 | - | 12 | 1790 | |||

Pdcd4
Genomic Map