siRNA Details
| Gene Symbol: Padi4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011061.2 | NP_035191.2 | 2, 3 | 304 | ||
| GenBank mRNA | AB013850.1 | - | 2, 3 | 297 | ||
| AK137625.1 | - | 2, 3 | 305 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Padi4| Gene Symbol: Padi4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011061.2 | NP_035191.2 | 2, 3 | 304 | ||
| GenBank mRNA | AB013850.1 | - | 2, 3 | 297 | ||
| AK137625.1 | - | 2, 3 | 305 | |||