siRNA Details
| Gene Symbol: Pex11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011069.3 | NP_035199.3 | 4 | 1407 | ||
| GenBank mRNA | AA462527.1 | - | 1 | 388 | ||
| AK005236.1 | - | 4 | 1154 | |||
| AK011709.1 | - | 4 | 1205 | |||
| AK082778.1 | - | 4 | 1127 | |||
| BC013812.1 | - | 4 | 1156 | 3483892 | ||
| BC054414.1 | - | 4 | 1155 | 3257217 | ||

Pex11b
Genomic Map