siRNA Details
| Gene Symbol: Itga3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC053031.1 | - | 4 | 1003 | 6401146 | |
| BC062205.1 | - | 3 | 778 | 4950774 | ||
| BC096741.1 | - | 5 | 981 | |||
| D13867.1 | - | 4 | 625 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itga3| Gene Symbol: Itga3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC053031.1 | - | 4 | 1003 | 6401146 | |
| BC062205.1 | - | 3 | 778 | 4950774 | ||
| BC096741.1 | - | 5 | 981 | |||
| D13867.1 | - | 4 | 625 | |||